missing translation for 'onlineSavingsMsg'
Learn More

Dirhenium decacarbonyl, sublimed

Catalog No. AC223990050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC223990050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC223990050 Supplier Thermo Scientific Chemicals Supplier No. 223990050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14285-68-8 | C10O10Re2 | 652.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

CAS 14285-68-8
Melting Point 170.0°C
Color Green-Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C10O10Re2
Linear Formula Re2(CO)10
Quantity 5 g
MDL Number MFCD00011198
Synonym carbon monoxide; rhenium 2+, decacarbonyldirhenium 250mg
Solubility Information Solubility in water: insoluble.
InChI Key ZIZHEHXAMPQGEK-UHFFFAOYSA-N
SMILES [O]#C[Re](C#[O])(C#[O])(C#[O])(C#[O])[Re](C#[O])(C#[O])(C#[O])(C#[O])C#[O]
IUPAC Name carbon monoxide;rhenium
Molecular Weight (g/mol) 652.51
PubChem CID 498777
Formula Weight 652.51
Physical Form Crystals
Chemical Name or Material Dirhenium decacarbonyl
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if inhaled.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
EINECSNumber 238-202-8
TSCA TSCA

RUO – Research Use Only