missing translation for 'onlineSavingsMsg'
Learn More

Direct Yellow 8, TCI America™

Catalog No. D111325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D111325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D111325G Supplier TCI America Supplier No. D111325G
Only null left

Chemical Identifiers

CAS 10130-29-7
Molecular Formula C24H19N4NaO5S2
Molecular Weight (g/mol) 530.55
MDL Number MFCD00005779
InChI Key OZRXNTRXLHTVPI-UHFFFAOYSA-M
Synonym Acid Yellow 186, Coomassie Yellow 6G, Direct Pure Yellow 5G
PubChem CID 23668793
IUPAC Name sodium 6-methyl-2-(4-{2-[2-oxo-1-(phenylcarbamoyl)propylidene]hydrazin-1-yl}phenyl)-1,3-benzothiazole-7-sulfonate
SMILES [Na+].CC(=O)C(=NNC1=CC=C(C=C1)C1=NC2=CC=C(C)C(=C2S1)S([O-])(=O)=O)C(=O)NC1=CC=CC=C1

Specifications

Quantity 25 g
Formula Weight 530.55
Physical Form Crystalline Powder
Chemical Name or Material Direct Yellow 8
EINECSNumber (5)-1180
TSCA Yes