missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Direct TIBC Reagents

Catalog No. TS3500
Change view
Click to view available options
Description:
Direct TIBC Calibrator
Direct TIBC Reagent
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
TS3500 Direct TIBC Reagent
TS3000 Direct TIBC Calibrator
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. TS3500 Supplier Thermo Scientific™ Supplier No. TS3500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Decrease turnaround time with Thermo Scientific™ Direct TIBC Reagents, which deliver a fully automated direct measurement of total iron-binding capacity.

This assay streamlines laboratory testing by maximizing lab productivity, minimizing operator error, improving turnaround times and eliminating the need for sample splitting. From centrifuge to analyzer, no sample pre-treatment is necessary.
  • Increase accuracy of results by as much as 20% over UIBC+ Iron methodologies1
  • Excellent correlation with Alumina Column, Magnetic and Transferrin TIBC methodologies
  • Unaffected by lipemia, bilirubinemia, hemolysis and other potentially interfering compounds
  • Liquid stable, ready-to-use reagent
  • Applications available for a variety of chemistry analyzers
1Siek G, Lawlor J, Pelczar D, Sane M, Musto J. Direct Serum Total Iron-binding Capacity Assay Suitable for Automated Analyzers. Clinical Chemistry. 2002; 48(1):161-166

Specifications

Certifications/Compliance Not CE Marked
Content And Storage 2°C to 8°C
Description Direct TIBC Reagent
CE Marker No
Detectable Analytes Iron
Sample Type Serum
Type Reagent
Methods Compatibility Colorimetric/Endpoint
Quantity 2 x 30 mL (R1); 2 x 15 mL (R2)
Stability 18 months
State at Room Temperature Liquid
Form Liquid
Show More Show Less