missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Direct Thermal Cryogenic Storage Rectangular Labels
GREENER_CHOICE

DYMO compatable storage labels designed to withstand extreme cold temperatures as low as -196°C.

Supplier:  Fisherbrand™ FISHDTC2

 View more versions of this product

Catalog No. 11010281


Only null left

Description

Description

  • Withstands temperatures from -196°C up to +80°C
  • Compatible with DYMO label printers and other direct thermal printers
  • Various sizes, colors, and configurations available
Specifications

Chemical Identifiers

Permanent
Cryogenic Label
English
Rectangle with Rounded Corners
Roll
Non-sterile
Yes

Specifications

Permanent
Matte
Cryogenic Label
English
Rectangle with Rounded Corners
Blue
Roll
Non-sterile
Yes
27 x 13 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions