missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Direct Scarlet B, TCI America™
Specifications
Specifications
| Quantity | 25 g |
| Formula Weight | 676.63 |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Direct Scarlet B |
Safety and Handling
| EINECSNumber | (5)-1239 |
| TSCA | Yes |