missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Direct Blue 86, TCI America™
Supplier: TCI America C111825G
Specifications
| Direct Blue 86 | |
| Blue-Purple | |
| MFCD00070592 | |
| LSASILAMYYWGBH-UHFFFAOYSA-L | |
| 780.16 | |
| 780.16 |
| 1330-38-7 | |
| C32H14CuN8Na2O6S2 | |
| 25 g | |
| C1=CC=C2C(=C1)C3=NC4=C5C=CC(=CC5=C([N-]4)N=C6C7=CC=CC=C7C(=N6)N=C8C9=C(C=C(C=C9)S(=O)(=O)[O-])C(=NC2=N3)[N-]8)S(=O)(=O)[O-].[Na+].[Na+].[Cu+2] | |
| 76045758 | |
| Crystalline Powder |
Chemical Identifiers
| 1330-38-7 | |
| 780.16 | |
| LSASILAMYYWGBH-UHFFFAOYSA-L | |
| C1=CC=C2C(=C1)C3=NC4=C5C=CC(=CC5=C([N-]4)N=C6C7=CC=CC=C7C(=N6)N=C8C9=C(C=C(C=C9)S(=O)(=O)[O-])C(=NC2=N3)[N-]8)S(=O)(=O)[O-].[Na+].[Na+].[Cu+2] |
| C32H14CuN8Na2O6S2 | |
| MFCD00070592 | |
| 76045758 |
Safety and Handling
EINECSNumber : (5)-1298
RTECSNumber : GS2169225
TSCA : Yes