missing translation for 'onlineSavingsMsg'
Learn More

Direct Blue 14, TCI America™

Catalog No. T055625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T055625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T055625G Supplier TCI America Supplier No. T055625G

Chemical Identifiers

CAS 72-57-1
Molecular Formula C34H24N6Na4O14S4
Molecular Weight (g/mol) 960.79
MDL Number MFCD00003969
InChI Key WMYVHJWZUUEZNE-ARWFNKCKSA-J
Synonym Trypan Blue
PubChem CID 9562061
IUPAC Name tetrasodium (3Z)-5-amino-3-[2-(4'-{2-[(2Z)-8-amino-1-oxo-3,6-disulfonato-1,2-dihydronaphthalen-2-ylidene]hydrazin-1-yl}-3,3'-dimethyl-[1,1'-biphenyl]-4-yl)hydrazin-1-ylidene]-4-oxo-3,4-dihydronaphthalene-2,7-disulfonate
SMILES [Na+].[Na+].[Na+].[Na+].CC1=CC(=CC=C1N\N=C1\C(=O)C2=C(N)C=C(C=C2C=C1S([O-])(=O)=O)S([O-])(=O)=O)C1=CC=C(N\N=C2\C(=O)C3=C(N)C=C(C=C3C=C2S([O-])(=O)=O)S([O-])(=O)=O)C(C)=C1

Specifications

Formula Weight 960.79
Color Black/Green
Quantity 25 g
Physical Form Crystal/Powder
Chemical Name or Material Direct Blue 14
EINECSNumber (5)-1285
RTECSNumber QJ6475000
TSCA Yes