missing translation for 'onlineSavingsMsg'
Learn More

Dipropylmalonic Acid 99.0+%, TCI America™

Catalog No. D09341G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D09341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D09341G Supplier TCI America Supplier No. D09341G

Chemical Identifiers

CAS 1636-27-7
Molecular Formula C9H16O4
Molecular Weight (g/mol) 188.223
MDL Number MFCD00051476
InChI Key DIRSQLKNZQKDBK-UHFFFAOYSA-N
Synonym dipropylmalonic acid, 2,2-dipropylmalonic acid, propanedioic acid, dipropyl, di-n-propylmalonic acid, malonic acid, dipropyl, 4,4-heptanedicarboxylic acid, heptane-4,4-dicarboxylic acid, dipropylpropanedioic acid, dipropylmalonsaure, dipropylmalonic aicd
PubChem CID 74221
IUPAC Name 2,2-dipropylpropanedioic acid
SMILES CCCC(CCC)(C(=O)O)C(=O)O

Specifications

Melting Point 158°C
Color White
Quantity 1 g
Formula Weight 188.22
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dipropylmalonic Acid