missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dipropylene glycol monobutyl ether
CAS: 29911-28-2 | C10H22O3 | 190.28 g/mol
Supplier: Thermo Scientific Chemicals 467260025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Dipropylene glycol monobutyl ether | |
| -75.0°C | |
| 230.0°C | |
| C10H22O3 | |
| 0.91 | |
| CUVLMZNMSPJDON-UHFFFAOYNA-N | |
| 1-[(1-butoxypropan-2-yl)oxy]propan-2-ol | |
| 4.35 mPa.s (25°C) |
| 29911-28-2 | |
| 0.9100 g/mL | |
| 100°C | |
| 2.5 L | |
| Solubility aq. soln.: 40-45 g/L (25°C). | |
| CCCCOCC(C)OCC(C)O | |
| 190.28 | |
| 190.28 |
Chemical Identifiers
| 29911-28-2 | |
| 190.28 | |
| 1-[(1-butoxypropan-2-yl)oxy]propan-2-ol |
| C10H22O3 | |
| CUVLMZNMSPJDON-UHFFFAOYNA-N | |
| CCCCOCC(C)OCC(C)O |
Safety and Handling
EINECSNumber : 249-951-5
RUO – Research Use Only