missing translation for 'onlineSavingsMsg'
Learn More

Dipropylene glycol monobutyl ether

Catalog No. AC467260010
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC467260010 1 L
AC467260025 2.5 L
AC467260100 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC467260010 Supplier Thermo Scientific Chemicals Supplier No. 467260010

CAS: 29911-28-2 | C10H22O3 | 190.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 29911-28-2
Molecular Formula C10H22O3
Molecular Weight (g/mol) 190.28
InChI Key CUVLMZNMSPJDON-UHFFFAOYNA-N
IUPAC Name 1-[(1-butoxypropan-2-yl)oxy]propan-2-ol
SMILES CCCCOCC(C)OCC(C)O

Specifications

Melting Point -75.0°C
Density 0.9100 g/mL
Boiling Point 230.0°C
Flash Point 100°C
Quantity 1 L
Specific Gravity 0.91
Solubility Information Solubility aq. soln.: 40-45 g/L (25°C).
Viscosity 4.35 mPa.s (25°C)
Formula Weight 190.28
Chemical Name or Material Dipropylene glycol monobutyl ether
EINECSNumber 249-951-5

RUO – Research Use Only