missing translation for 'onlineSavingsMsg'
Learn More

Dipropylene Glycol (mixture of isomers) 95.0+%, TCI America™

Catalog No. D093325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D093325G 25 g
D0933500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D093325G Supplier TCI America Supplier No. D093325G

Chemical Identifiers

CAS 25265-71-8
Molecular Formula C6H14O3
MDL Number MFCD00004538

Specifications

Color Colorless
Quantity 25 g
Formula Weight 134.18
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Dipropylene Glycol (mixture of isomers)
EINECSNumber (2)-0413
RTECSNumber UB8765000
TSCA Yes