Learn More
Dipropylene glycol, mixture of isomers, 99%
CAS: 25265-71-8 | C6H14O3 | 134.18 g/mol
$76.07 - $213.60
Chemical Identifiers
| CAS | 25265-71-8 |
|---|---|
| Molecular Weight (g/mol) | 134.18 |
| InChI Key | AZUXKVXMJOIAOF-UHFFFAOYNA-N |
| PubChem CID | 8087 |
| IUPAC Name | 1-(2-hydroxypropoxy)propan-2-ol |
| SMILES | CC(O)COCC(C)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1871636
|
Thermo Scientific Chemicals
A1871636 |
500 g |
Each for $76.07
|
|
|||||
|
AAA187160E
|
Thermo Scientific Chemicals
A187160E |
2500 g |
Each for $213.60
|
|
|||||
Description
Dipropylene glycol, mixture of isomers is used as a solvent, plasticizer, polymerization initiator and monomer. It acts as an additive used in perfumes, skin and hair care products. It is an active ingredient in commercial fog fluid, which is used as an entertainment in fog machines. Further, it is used as an intermediate in polyurethane polyols, alkyd resins and non-reinforced polyesters. In addition to this, it is used in dyes, paints, hydraulic brake fluids and cutting oils.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDipropylene glycol, mixture of isomers is used as a solvent, plasticizer, polymerization initiator and monomer. It acts as an additive used in perfumes, skin and hair care products. It is an active ingredient in commercial fog fluid, which is used as an entertainment in fog machines. Further, it is used as an intermediate in polyurethane polyols, alkyd resins and non-reinforced polyesters. In addition to this, it is used in dyes, paints, hydraulic brake fluids and cutting oils.
Solubility
Miscible with cold water, hot water, methanol, diethyl ether and ethanol.
Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Chemical Identifiers
| 25265-71-8 | |
| AZUXKVXMJOIAOF-UHFFFAOYNA-N | |
| 1-(2-hydroxypropoxy)propan-2-ol |
| 134.18 | |
| 8087 | |
| CC(O)COCC(C)O |
Specifications
| 25265-71-8 | |
| 1.023 | |
| 0.99 | |
| 1.441 | |
| 500 g | |
| Miscible with cold water,hot water,methanol,diethyl ether and ethanol. | |
| CC(O)COCC(C)O | |
| 134.18 | |
| 134.18 |
| °C to 32°C | |
| 231°C to 235°C | |
| C6H14O3 | |
| MFCD00004538 | |
| 1,1'-oxydi-2-propanol, 2-propanol, 1,1'-oxybis, 1,1'-oxydipropan-2-ol, bis 2-hydroxypropyl ether, 4-oxaheptane-2,6-diol, 1,1'-oxybis-2-propanol, 2,2'-dihydroxydipropyl ether, 2-propanol, 1,1'-oxydi, niax catalyst d-19, 1,1'-dimethyldiethylene glycol | |
| AZUXKVXMJOIAOF-UHFFFAOYNA-N | |
| 1-(2-hydroxypropoxy)propan-2-ol | |
| 8087 | |
| Dipropylene glycol, mixture of isomers |
Safety and Handling
EINECSNumber : 246-770-3
RTECSNumber : UB9765000
TSCA : Yes
storageNote1 : Hygroscopic
Recommended Storage : Ambient temperatures
RUO – Research Use Only