missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dipropylene Glycol Methyl Ether Acetate, 98%, Spectrum™ Chemical
Specifications
Specifications
| Acidity | 0.0005 |
| CAS Max % | 1 |
| Packaging | Steel Can |
| Quantity | 20 L |
| Percent Purity | 0.98 |
| Grade | Reagent |
CAUTION: For use by qualified personnel for research and development or other authorized use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.