missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dipropylene Glycol Dimethyl Ether (mixture of isomers) 94.0+%, TCI America™
$41.02 - $87.60
Chemical Identifiers
| CAS | 111109-77-4 |
|---|---|
| Molecular Formula | C8H18O3 |
| MDL Number | MFCD00210047 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D428325ML
|
TCI America
D428325ML |
25 mL |
Each for $41.02
|
|
|||||
|
D4283500ML
|
TCI America
D4283500ML |
500 mL |
Each for $87.60
|
|
|||||
Chemical Identifiers
| 111109-77-4 | |
| C8H18O3 | |
| MFCD00210047 |
Specifications
| 111109-77-4 | |
| C8H18O3 | |
| 25 mL | |
| ≥94.0% (GC) | |
| Dipropylene Glycol Dimethyl Ether (mixture of isomers) |
| Colorless | |
| MFCD00210047 | |
| 162.23 | |
| Liquid |
Safety and Handling
EINECSNumber : (7)-1321
RTECSNumber : TZ5487500
TSCA : Yes