missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dipropylene Glycol Dimethyl Ether (mixture of isomers) 94.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Quantity | 25 mL |
| Formula Weight | 162.23 |
| Percent Purity | ≥94.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Dipropylene Glycol Dimethyl Ether (mixture of isomers) |
Safety and Handling
| EINECSNumber | (7)-1321 |
| RTECSNumber | TZ5487500 |
| TSCA | Yes |