missing translation for 'onlineSavingsMsg'
Learn More

Dipropylene Glycol Diacrylate (stabilized with MEHQ) 80.0+%, TCI America™

Catalog No. p-7127745
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D347025G 25 g
D3470500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D347025G Supplier TCI America Supplier No. D347025G
Only null left

Chemical Identifiers

CAS 57472-68-1
Molecular Formula C12H18O5

Specifications

Color Yellow
Quantity 25 g
Formula Weight 242.27
Percent Purity ≥80.0% (GC)
Physical Form Liquid
Chemical Name or Material Dipropylene Glycol Diacrylate (stabilized with MEHQ)
EINECSNumber (7)-0152
RTECSNumber UD3643950
TSCA Yes