missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dipropylene Glycol, 99%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor D1207200LTBL
Specifications
| 0.0001 | |
| 1 | |
| C6H14O3 | |
| QYSXYAURTRCDJU-UHFFFAOYNA-N | |
| 1-(1-hydroxypropoxy)propan-1-ol | |
| 0.99 |
| 25265-71-8 | |
| Drum | |
| 200 L | |
| CCC(O)OC(O)CC | |
| 134.18 | |
| Reagent |
Chemical Identifiers
| 25265-71-8 | |
| 134.18 | |
| 1-(1-hydroxypropoxy)propan-1-ol |
| C6H14O3 | |
| QYSXYAURTRCDJU-UHFFFAOYNA-N | |
| CCC(O)OC(O)CC |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.