missing translation for 'onlineSavingsMsg'
Learn More

Dipropylene Glycol, 99%, Spectrum™ Chemical

Catalog No. 18610286
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610286 1 L Amber Glass Bottle
18610289 4 L Amber Glass Bottle
18610287 20 L Steel Can
18610288 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18610286 Supplier Spectrum Chemical Mfg Cor Supplier No. D12071LT

Chemical Identifiers

CAS 25265-71-8
Molecular Formula C6H14O3
Molecular Weight (g/mol) 134.18
InChI Key QYSXYAURTRCDJU-UHFFFAOYNA-N
IUPAC Name 1-(1-hydroxypropoxy)propan-1-ol
SMILES CCC(O)OC(O)CC

Specifications

Acidity 0.0001
CAS Max % 1
Packaging Amber Glass Bottle
Quantity 1 L
Percent Purity 0.99
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.