missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dipropyl 2,5-Pyridinedicarboxylate 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 175°C |
| Quantity | 5 g |
| Formula Weight | 251.28 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Dipropyl 2,5-Pyridinedicarboxylate |
Safety and Handling
| EINECSNumber | (5)-0746 |
| RTECSNumber | US8000000 |
| TSCA | No |