missing translation for 'onlineSavingsMsg'
Learn More

Dipropargylamine 97.0+%, TCI America™

Catalog No. D46855ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D46855ML 5 mL
D46851ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D46855ML Supplier TCI America Supplier No. D46855ML
Only null left

Chemical Identifiers

CAS 6921-28-4
Molecular Formula C6H7N
Molecular Weight (g/mol) 93.129
MDL Number MFCD00008574
InChI Key RGSODMOUXWISAG-UHFFFAOYSA-N
Synonym Di(2-propynyl)amine
PubChem CID 23350
IUPAC Name N-prop-2-ynylprop-2-yn-1-amine
SMILES C#CCNCC#C

Specifications

Boiling Point 65°C
Quantity 5 mL
UN Number 2734
Formula Weight 93.13
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material Dipropargylamine
RTECSNumber JM4551300
TSCA No