missing translation for 'onlineSavingsMsg'
Learn More

Dipotassium Dihydrogen Ethylenediaminetetraacetate Dihydrate 98.0+%, TCI America™

Catalog No. D3901500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3901500G 500 g
D390125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D3901500G Supplier TCI America Supplier No. D3901500G

Chemical Identifiers

CAS 25102-12-9
Molecular Formula C10H18K2N2O10
Molecular Weight (g/mol) 404.455
MDL Number MFCD00150036
InChI Key JFROQFOCFOKDKU-UHFFFAOYSA-L
Synonym ethylenediaminetetraacetic acid dipotassium salt dihydrate, dipotassium dihydrogen ethylenediaminetetraacetate dihydrate, potassium 2,2'-2-bis carboxymethyl amino ethyl azanediyl diacetate dihydrate, edta dipotassium salt dihydrate, edta dipotassium salt, ethylenediaminetetraacetic acid,dipotassium salt dihydrate, potassium ethylenediaminetetraacetate dibasic, dipotassium ethylenediaminetetraacetate dihydrate, ethylenedinitrilo tetraacetic acid dipotassium salt, acmc-209ggx
PubChem CID 53486451
IUPAC Name dipotassium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate
SMILES C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)O)CC(=O)O.O.O.[K+].[K+]

Specifications

Melting Point 272°C
Color White
Quantity 500 g
Formula Weight 368.42
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dipotassium Dihydrogen Ethylenediaminetetraacetate Dihydrate
EINECSNumber (2)-1265
TSCA Yes