missing translation for 'onlineSavingsMsg'
Learn More

(-)-Dipivaloyl-L-tartaric Acid 98.0+%, TCI America™

Catalog No. p-7127441
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26865G 5 g
D268625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D26865G Supplier TCI America Supplier No. D26865G
Only null left

Chemical Identifiers

CAS 65259-81-6
Molecular Formula C14H22O8
Molecular Weight (g/mol) 318.32
MDL Number MFCD00066535
InChI Key UFHJEZDFEHUYCR-YZYOREDDNA-N
PubChem CID 16212279
IUPAC Name (2R,3R)-2,3-bis[(2,2-dimethylpropanoyl)oxy]butanedioic acid
SMILES CC(C)(C)C(=O)O[C@H]([C@@H](OC(=O)C(C)(C)C)C(O)=O)C(O)=O

Specifications

Melting Point 150°C
Color White
Quantity 5 g
Synonym +-dipivaloyl-d-tartaric acid, 2s,3s-2,3-bis pivaloyloxy succinic acid, d-+-dipivaloyl tartaric acid, pubchem8173, --bis 2,2-dimethylpropionyl-l-tartaric acid, dipivaloyl-l-tartaric acid, +-dipivaloyl-d-tartaricacid, +-dpta, ?-o,o'-di-pivaloyl-l-tartaric acid, +-o,o'-di-pivaloyl-d-tartaric acid
Formula Weight 318.32
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (-)-Dipivaloyl-L-tartaric Acid