missing translation for 'onlineSavingsMsg'
Learn More

Diphosphoryl Chloride 98.0+%, TCI America™

Catalog No. D288325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D288325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D288325G Supplier TCI America Supplier No. D288325G

Chemical Identifiers

CAS 13498-14-1
Molecular Formula Cl4O3P2
Molecular Weight (g/mol) 251.745
MDL Number MFCD00061542
InChI Key CNTIXUGILVWVHR-UHFFFAOYSA-N
Synonym diphosphoryl chloride, pyrophosphoryl chloride, diphosphoryl tetrachloride, diphosphoric tetrachloride, pyrophosphoric tetrachloride, dichlorophosphoryl oxyphosphonoyl dichloride, pyrophosphoryl tetrachloride, pyrophosphorylchloride, phosphorodichloridic anhydride
PubChem CID 4432411
SMILES O=P(OP(=O)(Cl)Cl)(Cl)Cl

Specifications

Color Yellow
Physical Form Liquid
Quantity 25 g
UN Number 3264
Formula Weight 251.74
Percent Purity ≥98.0% (T)
Chemical Name or Material Diphosphoryl Chloride
EINECSNumber (1)-0245
TSCA No