missing translation for 'onlineSavingsMsg'
Learn More

Diphenyltin dichloride, 90+%

Catalog No. p-7080319
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA7113904 2 g
AA7113909 10 g
AA7113918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA7113904 Supplier Thermo Scientific Chemicals Supplier No. 07113904

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1135-99-5 | C12H12Cl2Sn | 345.84 g/mol

Used in the manufacture of chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1135-99-5
Molecular Formula C12H12Cl2Sn
Molecular Weight (g/mol) 345.84
MDL Number MFCD00000516
InChI Key WSXPROXKZIMZIU-UHFFFAOYSA-N
Synonym diphenyltin dichloride, stannane, dichlorodiphenyl, dichlorodiphenyltin, diphenyldichlorotin, diphenyltin chloride, diphenylstannyl dichloride, tin, diphenyl-, dichloride, dichloro diphenyl stannane, diphenylstannium dichloride, unii-0y7mz4k0dr
PubChem CID 14342
IUPAC Name dichloro(diphenyl)stannane
SMILES Cl.Cl.[SnH2](C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 41°C to 43°C
Color White
Boiling Point 333°C to 337°C (decomposition)
Flash Point >150°C (302°F)
Linear Formula (C6H5)2SnCl2
Quantity 2 g
Sensitivity Moisture sensitive
Solubility Information Sparingly Soluble (2.4E-3g/L) (25°C),Calc.
Formula Weight 343.81
Percent Purity 95%
Physical Form Crystalline
Chemical Name or Material Diphenyltin dichloride
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 214-496-3
RTECSNumber WH7253000
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only