missing translation for 'onlineSavingsMsg'
Learn More

Diphenyltetramethyldisilane, 97%

Catalog No. AC387880010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC387880010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC387880010 Supplier Thermo Scientific Chemicals Supplier No. 387880010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1145-98-8 | C16H22Si2 | 270.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1145-98-8
Molecular Formula C16H22Si2
Molecular Weight (g/mol) 270.32
MDL Number MFCD00054786
InChI Key IIOOIYHUTINYQO-UHFFFAOYSA-N
Synonym 1,2-diphenyltetramethyldisilane, 1,1,2,2-tetramethyl-1,2-diphenyldisilane, disilane,1,1,2,2-tetramethyl-1,2-diphenyl, 1,2-diphenyl-1,1,2,2-tetramethyldisilane, tetramethyldiphenyldisilan, amtsi083, tetramethyldisilane-1,2-diyldibenzene, bisphenyl-1,1,2,2-tetramethyldisilane, dimethyl phenyl silyl-dimethyl-phenylsilane, 1,1,2,2-tetramethyldiphenyldisilane
PubChem CID 136916
IUPAC Name [dimethyl(phenyl)silyl]-dimethyl-phenylsilane
SMILES C[Si](C)(C1=CC=CC=C1)[Si](C)(C)C2=CC=CC=C2

Specifications

Melting Point 33°C to 38°C
Color White to Brown
Boiling Point 145°C to 148°C
Flash Point 105°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula [C6H5Si(CH3)2]2
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 270.32
Percent Purity 97%
Physical Form Low Melting Mass
Chemical Name or Material Diphenyltetramethyldisilane
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only