missing translation for 'onlineSavingsMsg'
Learn More

Diphenylsilane 98.0+%, TCI America™

Catalog No. p-7127517
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28205G 5 g
D282025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D28205G Supplier TCI America Supplier No. D28205G
Only null left

Chemical Identifiers

CAS 775-12-2
Molecular Formula C12H10Si
Molecular Weight (g/mol) 182.297
MDL Number MFCD00003002
InChI Key BPYFPNZHLXDIGA-UHFFFAOYSA-N
Synonym diphenylsilane, silane, diphenyl, diphenylsilicon, benzene, 1,1'-silylenebis, diphenyl silane, di phenyl silicon, diphenylsilylene radical, ph 2sih2
PubChem CID 6327659
IUPAC Name cyclohexa-2,5-dien-1-ylidene(phenyl)silanide
SMILES C1=CC=C(C=C1)[Si-]=C2C=C[CH+]C=C2

Specifications

Color Yellow
Boiling Point 97°C
Quantity 5 g
Formula Weight 184.31
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diphenylsilane