missing translation for 'onlineSavingsMsg'
Learn More

Diphenylpropylphosphine 98.0+%, TCI America™

Supplier:  TCI America D26305G

Catalog No. D26305G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Diphenylpropylphosphine
Colorless
C15H17P
5 g
AAXGWYDSLJUQLN-UHFFFAOYSA-N
diphenyl(propyl)phosphane
82100
≥98.0% (GC)
7650-84-2
165°C
MFCD00015216
diphenyl-n-propylphosphine, phosphine, diphenylpropyl, propyldiphenylphosphine, n-propyldiphenylphosphine, diphenyl propyl phosphine, diphenylphosphinopropane, diphenyl propyl phosphane, diphenylphospinopropane, di phenyl-propylphosphane
CCCP(C1=CC=CC=C1)C1=CC=CC=C1
228.28
228.27
Liquid

Chemical Identifiers

7650-84-2
228.28
AAXGWYDSLJUQLN-UHFFFAOYSA-N
82100
CCCP(C1=CC=CC=C1)C1=CC=CC=C1
C15H17P
MFCD00015216
diphenyl-n-propylphosphine, phosphine, diphenylpropyl, propyldiphenylphosphine, n-propyldiphenylphosphine, diphenyl propyl phosphine, diphenylphosphinopropane, diphenyl propyl phosphane, diphenylphospinopropane, di phenyl-propylphosphane
diphenyl(propyl)phosphane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : SY9150000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions