missing translation for 'onlineSavingsMsg'
Learn More

Diphenylphosphinic Chloride 98.0+%, TCI America™

Catalog No. C141525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C141525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C141525G Supplier TCI America Supplier No. C141525G
Only null left

Specifications

CAS 1499-21-4
Color Yellow
Boiling Point 222°C
Molecular Formula C12H10ClOP
Quantity 25 g
MDL Number MFCD00002077
UN Number 2920
Synonym diphenylphosphinic chloride, diphenylphosphinyl chloride, phosphinic chloride, diphenyl, diphenylphosphinoyl chloride, chlorodiphenylphosphine oxide, dpp-cl, unii-5gp30gy04v, diphenyl phosphinic chloride, diphenylphosphinochloridic acid, diphenylphosphinic acid chloride
InChI Key QPQGTZMAQRXCJW-UHFFFAOYSA-N
SMILES ClP(=O)(C1=CC=CC=C1)C1=CC=CC=C1
IUPAC Name diphenylphosphinoyl chloride
Molecular Weight (g/mol) 236.63
PubChem CID 73910
Formula Weight 236.63
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Diphenylphosphinic Chloride
Show More Show Less