Learn More
Diphenylphosphinic chloride, 97+%
CAS: 1499-21-4 | C12H10ClOP | 236.63 g/mol
Supplier: Thermo Scientific Chemicals A1172114
Description
Diphenylphosphinic chloride is a reagent that is used in the synthesis of bidentate ligands and peptide coupling agents. It is used as flame retardants. It also act as pharmaceutical adjuvant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDiphenylphosphinic chloride is a reagent that is used in the synthesis of bidentate ligands and peptide coupling agents. It is used as flame retardants. It also act as pharmaceutical adjuvant.
Solubility
Decomposes with water.
Notes
Moisture Sensitive. Store away from strong bases, moisture. Incompatible with oxidizing agents,, Amines, Alcohols.
Specifications
| Diphenylphosphinic chloride | |
| ∼20°C | |
| 210°C to 212°C (8 mmHg) | |
| C12H10ClOP | |
| MFCD00002077 | |
| UN3265 | |
| diphenylphosphinic chloride, diphenylphosphinyl chloride, phosphinic chloride, diphenyl, diphenylphosphinoyl chloride, chlorodiphenylphosphine oxide, dpp-cl, unii-5gp30gy04v, diphenyl phosphinic chloride, diphenylphosphinochloridic acid, diphenylphosphinic acid chloride | |
| QPQGTZMAQRXCJW-UHFFFAOYSA-N | |
| [chloro(phenyl)phosphoryl]benzene | |
| 73910 | |
| ≥97% |
| 1499-21-4 | |
| 1.242 | |
| >110°C (230°F) | |
| 1.609 | |
| 25 g | |
| Moisture sensitive | |
| Decomposes with water. | |
| ClP(=O)(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 236.63 | |
| 236.64 |
Chemical Identifiers
| 1499-21-4 | |
| 236.63 | |
| QPQGTZMAQRXCJW-UHFFFAOYSA-N | |
| 73910 | |
| ClP(=O)(C1=CC=CC=C1)C1=CC=CC=C1 |
| C12H10ClOP | |
| MFCD00002077 | |
| diphenylphosphinic chloride, diphenylphosphinyl chloride, phosphinic chloride, diphenyl, diphenylphosphinoyl chloride, chlorodiphenylphosphine oxide, dpp-cl, unii-5gp30gy04v, diphenyl phosphinic chloride, diphenylphosphinochloridic acid, diphenylphosphinic acid chloride | |
| [chloro(phenyl)phosphoryl]benzene |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H226-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 216-107-2
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only