missing translation for 'onlineSavingsMsg'
Learn More

Diphenylphosphinic Acid 98.0+%, TCI America™

Catalog No. p-7127832
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36275G 5 g
D362725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D36275G Supplier TCI America Supplier No. D36275G
Only null left

Chemical Identifiers

CAS 1707-03-5
Molecular Formula C12H11O2P
Molecular Weight (g/mol) 218.19
MDL Number MFCD00002132
InChI Key BEQVQKJCLJBTKZ-UHFFFAOYSA-N
Synonym phosphinic acid, diphenyl, hydroxydiphenylphosphine oxide, diphenylphosphinicacid, diphenyl phosphinic acid, diphenyl-phosphinic acid, acmc-1boli, di phenyl phosphinic acid, p,p-diphenylphosphinic acid, ksc180o2h, phosphinicacid, p,p-diphenyl
PubChem CID 15567
ChEBI CHEBI:37832
IUPAC Name diphenylphosphinic acid
SMILES OP(=O)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 195°C
Color White-Yellow
Quantity 5 g
Formula Weight 218.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Diphenylphosphinic Acid
RTECSNumber SZ5315000
TSCA No