missing translation for 'onlineSavingsMsg'
Learn More

Diphenylphosphinamide, 98%

Catalog No. p-7023269
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1665706 5 g
AAA1665714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1665706 Supplier Thermo Scientific Chemicals Supplier No. A1665706

CAS: 5994-87-6 | C12H12NOP | 217.21 g/mol

Diphenylphosphinamide is used to furnish 5H-oxazol-4-ones with diastereoselectivity and enantioselectivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenylphosphinamide is used to furnish 5H-oxazol-4-ones with diastereoselectivity and enantioselectivity.

Solubility
Soluble in chloroform, nitromethane, nitrobenzene.

Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 5994-87-6
Molecular Formula C12H12NOP
Molecular Weight (g/mol) 217.21
MDL Number MFCD00014766
InChI Key RIGIWEGXTTUCIQ-UHFFFAOYSA-N
Synonym diphenylphosphinamide, p,p-diphenylphosphinic amide, amino phenyl phosphoryl benzene, amino phenyl phosphoroso benzene, diphenyl phosphinic acid amide, azanyl phenyl phosphoryl benzene
PubChem CID 853560
SMILES NP(=O)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 160°C to 162°C
Quantity 5 g
Solubility Information Soluble in chloroform,nitromethane,nitrobenzene.
IUPAC Name [amino(phenyl)phosphoroso]benzene
Formula Weight 217.21
Percent Purity 98%
Chemical Name or Material Diphenylphosphinamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only