missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ α,α-Diphenylglycine, 98%
$155.66 - $1,525.08
Chemical Identifiers
| CAS | 3060-50-2 |
|---|---|
| Molecular Formula | C14H13NO2 |
| Molecular Weight (g/mol) | 227.26 |
| MDL Number | MFCD00008048 |
| InChI Key | YBONNYNNFBAKLI-UHFFFAOYSA-N |
| Synonym | 2,2-diphenylglycine, alpha,alpha-diphenylglycine, alpha-aminodiphenylacetic acid, glycine, 2,2-diphenyl, alpha-amino-alpha-phenylbenzeneacetic acid, .alpha.-aminodiphenylacetic acid, .alpha.,.alpha.-diphenylglycine, benzeneacetic acid, .alpha.-amino-.alpha.-phenyl, aminodiphenylacetic acid, phenytoin impurity c |
| PubChem CID | 18289 |
| IUPAC Name | 2-amino-2,2-diphenylacetic acid |
| SMILES | NC(C(O)=O)(C1=CC=CC=C1)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC171720050
|
Thermo Scientific™
171720050 |
5g | Glass bottle |
Each for $155.66
|
|
||||
|
AC171720250
|
Thermo Scientific™
171720250 |
25g | Glass bottle |
Each for $517.14
|
|
||||
|
AC171721000
|
Thermo Scientific™
171721000 |
100g | Glass bottle |
Each for $1,525.08
|
|
||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C14H13NO2 | |
| MFCD00008048 | |
| 2,2-diphenylglycine, alpha,alpha-diphenylglycine, alpha-aminodiphenylacetic acid, glycine, 2,2-diphenyl, alpha-amino-alpha-phenylbenzeneacetic acid, .alpha.-aminodiphenylacetic acid, .alpha.,.alpha.-diphenylglycine, benzeneacetic acid, .alpha.-amino-.alpha.-phenyl, aminodiphenylacetic acid, phenytoin impurity c | |
| 2-amino-2,2-diphenylacetic acid |
Specifications
| 3060-50-2 | |
| Authentic | |
| C14H13NO2 | |
| MFCD00008048 | |
| 2,2-diphenylglycine, alpha,alpha-diphenylglycine, alpha-aminodiphenylacetic acid, glycine, 2,2-diphenyl, alpha-amino-alpha-phenylbenzeneacetic acid, .alpha.-aminodiphenylacetic acid, .alpha.,.alpha.-diphenylglycine, benzeneacetic acid, .alpha.-amino-.alpha.-phenyl, aminodiphenylacetic acid, phenytoin impurity c | |
| NC(C(O)=O)(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 227.26 | |
| 227.26 | |
| α,α-Diphenylglycine |
| 1% max. | |
| 98% | |
| H2NC(C6H5)2CO2H | |
| 14,I,625 | |
| YBONNYNNFBAKLI-UHFFFAOYSA-N | |
| 2-amino-2,2-diphenylacetic acid | |
| 18289 | |
| 98% |
Safety and Handling
EINECSNumber : 221-299-6
RUO – Research Use Only