missing translation for 'onlineSavingsMsg'
Learn More

Diphenyleneiodonium chloride

Catalog No. p-7042984
Change view
Click to view available options
Quantity:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64838MF 50 mg
AAJ64838MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ64838MF Supplier Thermo Scientific Chemicals Supplier No. J64838MF
Only null left

CAS: 4673-26-1 | C12H8ClI | 314.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4673-26-1
Molecular Formula C12H8ClI
Molecular Weight (g/mol) 314.55
MDL Number MFCD00214165
InChI Key FCFZKAVCDNTYID-UHFFFAOYSA-M
Synonym diphenyleneiodonium chloride, dibenziodolium chloride, unii-7m9d81yz2n, dpi, 8? 3-iodatricyclo 7.4.0.0 2 ,? trideca-1 13 ,2,4,6,9,11-hexaen-8-ylium chloride, dibenziodolium, chloride, d0n0gj, c12h8i.cl, dibenzo b,d iodolium chloride, 2,2'-biphenylyleneiodonium chloride
PubChem CID 2733504
ChEBI CHEBI:77967
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3[I+]2.[Cl-]

Specifications

Melting Point >310°C
Color White
Quantity 50 mg
Formula Weight 314.55
Physical Form Powder
Chemical Name or Material Diphenyleneiodonium chloride
Hazard Category H319
Precautionary Statement P264b-P280i-P305+P351+P338
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only