missing translation for 'onlineSavingsMsg'
Learn More

Diphenylcarbazone (contains Diphenylcarbazide) 40.0+%, TCI America™

Catalog No. d088125g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D088125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D088125G Supplier TCI America Supplier No. D088125G
Only null left

Chemical Identifiers

CAS 538-62-5
Molecular Formula C13H12N4O
Molecular Weight (g/mol) 240.266
MDL Number MFCD00003024
InChI Key ZFWAHZCOKGWUIT-UHFFFAOYSA-N
PubChem CID 10860
IUPAC Name 1-anilino-3-phenyliminourea
SMILES C1=CC=C(C=C1)NNC(=O)N=NC2=CC=CC=C2

Specifications

Melting Point 157°C
Formula Weight 240.27
Percent Purity ≥40.0% (HPLC)
Quantity 25 g
Physical Form Crystal/Powder
Chemical Name or Material Diphenylcarbazone (contains Diphenylcarbazide)
EINECSNumber (9)-0624
RTECSNumber LQ9420000
TSCA Yes