Learn More
Diphenylcarbamyl chloride, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 167145000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Diphenylcarbamyl chloride | |
| Authentic | |
| Glass bottle | |
| (C6H5)2NCOCl | |
| MFCD00000633 | |
| diphenylcarbamyl chloride, diphenylcarbamoyl chloride, carbamic chloride, diphenyl, diphenylcarbamic chloride, n,n-diphenylcarbamyl chloride, diphenylchloroformamide, carbamoyl chloride, diphenyl, carabamic chloride, diphenyl, chloroformic acid diphenylamide, carbamic chloride, n,n-diphenyl | |
| XNBKKRFABABBPM-UHFFFAOYSA-N | |
| N,N-diphenylcarbamoyl chloride | |
| 65741 | |
| 98% |
| 83-01-2 | |
| 97.5% min. (GC) | |
| C13H10ClNO | |
| 500g | |
| 15, 3351 | |
| Solubility in water: reacts. | |
| C1=CC=C(C=C1)N(C2=CC=CC=C2)C(=O)Cl | |
| 231.68 | |
| 231.68 |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES:
GHS Signal Word: Danger
EINECSNumber : 201-450-2
Promotions