missing translation for 'onlineSavingsMsg'
Learn More

Diphenylcarbamyl chloride, 98%

Catalog No. p-7026851
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2311614 25 g
AAB2311622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2311614 Supplier Thermo Scientific Chemicals Supplier No. B2311614

CAS: 83-01-2 | C13H10ClNO | 231.679 g/mol

Diphenylcarbamoyl chloride was used to isolate peptido-keratan sulphate fragments from bovine intervertebral discs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenylcarbamoyl chloride was used to isolate peptido-keratan sulphate fragments from bovine intervertebral discs.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from oxidizing agents, water/ moisture, bases.

Chemical Identifiers

CAS 83-01-2
Molecular Formula C13H10ClNO
Molecular Weight (g/mol) 231.679
MDL Number MFCD00000633
InChI Key XNBKKRFABABBPM-UHFFFAOYSA-N
Synonym diphenylcarbamyl chloride, diphenylcarbamoyl chloride, carbamic chloride, diphenyl, diphenylcarbamic chloride, n,n-diphenylcarbamyl chloride, diphenylchloroformamide, carbamoyl chloride, diphenyl, carabamic chloride, diphenyl, chloroformic acid diphenylamide, carbamic chloride, n,n-diphenyl
PubChem CID 65741
IUPAC Name N,N-diphenylcarbamoyl chloride
SMILES C1=CC=C(C=C1)N(C2=CC=CC=C2)C(=O)Cl

Specifications

Melting Point 82°C to 86°C
UN Number UN3261
Beilstein 515312
Solubility Information Reacts with water.
Quantity 25 g
Odor Irritating
Formula Weight 231.68
Sensitivity Moisture Sensitive
Percent Purity 98%
Chemical Name or Material Diphenylcarbamyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 201-450-2
RTECSNumber EY5065000
TSCA Yes

RUO – Research Use Only