missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphenylcarbamoyl Chloride 98.0+%, TCI America™
$104.38 - $305.70
Chemical Identifiers
| CAS | 83-01-2 |
|---|---|
| Molecular Formula | C13H10ClNO |
| Molecular Weight (g/mol) | 231.679 |
| MDL Number | MFCD00000633 |
| InChI Key | XNBKKRFABABBPM-UHFFFAOYSA-N |
| Synonym | diphenylcarbamyl chloride, diphenylcarbamoyl chloride, carbamic chloride, diphenyl, diphenylcarbamic chloride, n,n-diphenylcarbamyl chloride, diphenylchloroformamide, carbamoyl chloride, diphenyl, carabamic chloride, diphenyl, chloroformic acid diphenylamide, carbamic chloride, n,n-diphenyl |
| PubChem CID | 65741 |
| IUPAC Name | N,N-diphenylcarbamoyl chloride |
| SMILES | C1=CC=C(C=C1)N(C2=CC=CC=C2)C(=O)Cl |
Chemical Identifiers
| 83-01-2 | |
| 231.679 | |
| XNBKKRFABABBPM-UHFFFAOYSA-N | |
| 65741 | |
| C1=CC=C(C=C1)N(C2=CC=CC=C2)C(=O)Cl |
| C13H10ClNO | |
| MFCD00000633 | |
| diphenylcarbamyl chloride, diphenylcarbamoyl chloride, carbamic chloride, diphenyl, diphenylcarbamic chloride, n,n-diphenylcarbamyl chloride, diphenylchloroformamide, carbamoyl chloride, diphenyl, carabamic chloride, diphenyl, chloroformic acid diphenylamide, carbamic chloride, n,n-diphenyl | |
| N,N-diphenylcarbamoyl chloride |
Specifications
| 83°C | |
| Blue-White | |
| MFCD00000633 | |
| diphenylcarbamyl chloride, diphenylcarbamoyl chloride, carbamic chloride, diphenyl, diphenylcarbamic chloride, n,n-diphenylcarbamyl chloride, diphenylchloroformamide, carbamoyl chloride, diphenyl, carabamic chloride, diphenyl, chloroformic acid diphenylamide, carbamic chloride, n,n-diphenyl | |
| C1=CC=C(C=C1)N(C2=CC=CC=C2)C(=O)Cl | |
| 5 g | |
| 65741 | |
| ≥98.0% (GC,T) | |
| Diphenylcarbamoyl Chloride |
| 83-01-2 | |
| C13H10ClNO | |
| 3261 | |
| XNBKKRFABABBPM-UHFFFAOYSA-N | |
| N,N-diphenylcarbamoyl chloride | |
| 231.679 | |
| 231.68 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : EY5065000
TSCA : Yes