missing translation for 'onlineSavingsMsg'
Learn More

Diphenylammonium Trifluoromethanesulfonate 98.0+%, TCI America™

Catalog No. D36831G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36831G 1 g
D36835G 5 g
D368325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D36831G Supplier TCI America Supplier No. D36831G
Only null left

Chemical Identifiers

CAS 164411-06-7
Molecular Formula C13H12F3NO3S
Molecular Weight (g/mol) 319.298
InChI Key MGEGQAUINMTPGX-UHFFFAOYSA-N
Synonym DPAT
PubChem CID 11209371
IUPAC Name diphenylazanium;trifluoromethanesulfonate
SMILES C1=CC=C(C=C1)[NH2+]C2=CC=CC=C2.C(F)(F)(F)S(=O)(=O)[O-]

Specifications

Color White
Quantity 1 g
Formula Weight 319.30
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Diphenylammonium Trifluoromethanesulfonate