missing translation for 'onlineSavingsMsg'
Learn More

Diphenylamine Sulfate 98.0+%, TCI America™

Catalog No. D0875500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0875500G 500 g
D087525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0875500G Supplier TCI America Supplier No. D0875500G
Only null left

Chemical Identifiers

CAS 587-84-8
Molecular Formula C12H13NO4S
Molecular Weight (g/mol) 267.30
MDL Number MFCD00042016
InChI Key IPZMDJYHJNHGML-UHFFFAOYSA-N
Synonym diphenylamine sulfate, benzenamine, n-phenyl-, sulfate 1:1, usaf ek-743, n-phenylaniline; sulfuric acid, diphenylamine, hydrogen sulfate, diphenylammonium hydrogen sulphate, diphenylamine; sulfuric acid, diphenylaminesulfate, bis n-phenylanilinium sulphate, diphenylaminsulfat
PubChem CID 11486
IUPAC Name N-phenylaniline; sulfuric acid
SMILES OS(O)(=O)=O.N(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 150°C
Color White
Quantity 500 g
Formula Weight 267.30
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Diphenylamine Sulfate
RTECSNumber JJ9285000
TSCA Yes