missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphenylamine, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
$5,536.06 - $5,536.06
Chemical Identifiers
| CAS | 122-39-4 |
|---|---|
| Molecular Formula | C12H11N |
| Molecular Weight (g/mol) | 169.227 |
| InChI Key | DMBHHRLKUKUOEG-UHFFFAOYSA-N |
| Synonym | diphenylamine, n-phenylbenzenamine, anilinobenzene, benzenamine, n-phenyl, n,n-diphenylamine, phenylaniline, scaldip, n-phenylbenzeneamine, big dipper, no-scald |
| PubChem CID | 11487 |
| ChEBI | CHEBI:4640 |
| IUPAC Name | N-phenylaniline |
| SMILES | C1=CC=C(C=C1)NC2=CC=CC=C2 |
Chemical Identifiers
| 122-39-4 | |
| 169.227 | |
| diphenylamine, n-phenylbenzenamine, anilinobenzene, benzenamine, n-phenyl, n,n-diphenylamine, phenylaniline, scaldip, n-phenylbenzeneamine, big dipper, no-scald | |
| CHEBI:4640 | |
| C1=CC=C(C=C1)NC2=CC=CC=C2 |
| C12H11N | |
| DMBHHRLKUKUOEG-UHFFFAOYSA-N | |
| 11487 | |
| N-phenylaniline |
Specifications
| 122-39-4 | |
| Wide Mouth Amber Glass Bottle | |
| 500 g | |
| diphenylamine, n-phenylbenzenamine, anilinobenzene, benzenamine, n-phenyl, n,n-diphenylamine, phenylaniline, scaldip, n-phenylbenzeneamine, big dipper, no-scald | |
| C1=CC=C(C=C1)NC2=CC=CC=C2 | |
| 169.227 | |
| CHEBI:4640 | |
| BAKER ANALYZED™ ACS Reagent | |
| Diphenylamine |
| 150.00°C | |
| C12H11N | |
| 1L = 1.16kg | |
| DMBHHRLKUKUOEG-UHFFFAOYSA-N | |
| N-phenylaniline | |
| 11487 | |
| 169.23 | |
| Crystals |