missing translation for 'onlineSavingsMsg'
Learn More

Diphenylamine, ACS

Catalog No. p-7050367
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3661014 25 g
AA3661022 100 g
AA3661036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3661014 Supplier Thermo Scientific Chemicals Supplier No. 03661014
Only null left

CAS: 122-39-4 | C12H11N | 169.227 g/mol

Diphenylamine is employed as harvest scald inhibitor for apples indoor drench treatments due to its antioxidant property. Alkyl derivatives of diphenylamine are used as anti-ozonants in the rubber manufacturing. It is used as lubricants and acts as a stabilizer for plastics. In the analytical chemistry, it is used to test the nitrate ion in association with ammonium chloride in sulfuric acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 122-39-4
Molecular Formula C12H11N
Molecular Weight (g/mol) 169.227
MDL Number MFCD00003014
InChI Key DMBHHRLKUKUOEG-UHFFFAOYSA-N
Synonym diphenylamine, n-phenylbenzenamine, anilinobenzene, benzenamine, n-phenyl, n,n-diphenylamine, phenylaniline, scaldip, n-phenylbenzeneamine, big dipper, no-scald
PubChem CID 11487
ChEBI CHEBI:4640
IUPAC Name N-phenylaniline
SMILES C1=CC=C(C=C1)NC2=CC=CC=C2

Specifications

Melting Point 52°C to 55°C
Density 1.16 g/mL
Boiling Point 302°C
Flash Point 152°C (305°F)
Linear Formula (C6H5)2NH
Quantity 25 g
UN Number UN3077
Beilstein 508755
Sensitivity Air and light sensitive
Merck Index 14,3317
Solubility Information Soluble in ethanol,acetone,benzene,carbon tetrachloride,pyridine,ethyl acetate; Slightly soluble in chloroform.
Formula Weight 169.23
Grade ACS
Physical Form Solid
Chemical Name or Material Diphenylamine, For analysis ACS
Show More Show Less
Hazard Category H301+H311+H331-H373
Hazard Statement GHS H Statement
H301-H311-H331-H373
Precautionary Statement P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 204-539-4
RTECSNumber JJ7800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only