missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphenylamine, ACS Reagent Grade, Ricca Chemical
Specifications
Specifications
| CAS | 122-39-4 |
| CAS Min % | 98.0 |
| CAS Max % | 100.0 |
| Packaging | White Poly Wide Mouth Bottle |
| Molecular Formula | C12H11N |
| Quantity | 500 g |
| Synonym | diphenylamine, n-phenylbenzenamine, anilinobenzene, benzenamine, n-phenyl, n,n-diphenylamine, phenylaniline, scaldip, n-phenylbenzeneamine, big dipper, no-scald |
| InChI Key | DMBHHRLKUKUOEG-UHFFFAOYSA-N |
| SMILES | C1=CC=C(C=C1)NC2=CC=CC=C2 |
| IUPAC Name | N-phenylaniline |
| Show More |
Safety and Handling
| ShelfLife | 60 months |