missing translation for 'onlineSavingsMsg'
Learn More

Diphenylamine, ACS reagent

Catalog No. p-4187325
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC423650050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC423650050 Supplier Thermo Scientific Chemicals Supplier No. 423650050
Only null left

CAS: 122-39-4 | C12H11N | 169.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Chemical Identifiers

CAS 122-39-4
Molecular Formula C12H11N
Molecular Weight (g/mol) 169.23
MDL Number MFCD00003014
InChI Key DMBHHRLKUKUOEG-UHFFFAOYSA-N
Synonym diphenylamine, n-phenylbenzenamine, anilinobenzene, benzenamine, n-phenyl, n,n-diphenylamine, phenylaniline, scaldip, n-phenylbenzeneamine, big dipper, no-scald
PubChem CID 11487
ChEBI CHEBI:4640
IUPAC Name N-phenylaniline
SMILES C1=CC=C(C=C1)NC2=CC=CC=C2

Specifications

Melting Point 52.0°C to 54.0°C
Color White
Boiling Point 302.0°C
Flash Point 152°C
Ignition Residue 0.03% max.
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula (C6H5)2NH
Quantity 5 g
Beilstein 12, 174
Sensitivity (To nitrate) Passes Test
Merck Index 15, 3347
Solubility Information (in alcohol) Passes test
Formula Weight 169.23
Grade ACS Reagent
Physical Form Crystalline Flakes
Chemical Name or Material Diphenylamine, For analysis ACS
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if inhaled.
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Toxic if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 204-539-4

RUO – Research Use Only