missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphénylamine, réactif ACS
Description
Ce produit de marque Thermo Scientific Chemicals faisait à l’origine partie du portefeuille Acros Organics. Certains documents et informations d’étiquetage peuvent faire référence à l’ancienne marque. Le code produit/article (SKU) original d’Acros Organics n’a pas changé dans le cadre de la transition vers la marque Thermo Scientific Chemicals.
Indicateur
Spécifications
Spécifications
| Nom ou substance chimique | Diphenylamine |
| Nom Remarque | Pour analyse, ACS |
| Point de fusion | 52,0 °C à 54,0 °C |
| Couleur | Blanc |
| Point d’ébullition | 302.0 °C |
| Point d’éclair | 152 °C |
| Résidus de calcination | 0,03% max. |
| Spectre infrarouge | Authentique |
| Emballage | Bouteille en verre |
| Formule linéaire | (C6H5)2NH |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement: Toxic if inhaled. Toxic in contact with skin. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life with long lasting effects. Toxic if swallowed. |
| Precautionary Statement | GHS P Statement: Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Gently wash with plenty of soap and water. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Do not breathe dust/fume/gas/mist/vapors/spray. Avoid release to the environment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
| EINECSNumber | 204-539-4 |
RUO – Usage réservé à la recherche