missing translation for 'onlineSavingsMsg'
Learn More

Diphenylamine, ACS Grade, LabChem™

Catalog No. LC136109
Change view
Click to view available options
Quantity:
100 g
Packaging:
Amber Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC136109 100 g Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC136109 Supplier LabChem Supplier No. LC136109

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 122-39-4
Molecular Formula C12H11N
Molecular Weight (g/mol) 169.227
InChI Key DMBHHRLKUKUOEG-UHFFFAOYSA-N
Synonym diphenylamine, n-phenylbenzenamine, anilinobenzene, benzenamine, n-phenyl, n,n-diphenylamine, phenylaniline, scaldip, n-phenylbenzeneamine, big dipper, no-scald
PubChem CID 11487
ChEBI CHEBI:4640
IUPAC Name N-phenylaniline
SMILES C1=CC=C(C=C1)NC2=CC=CC=C2

Specifications

CAS Max % 100
Color White
Density 1.16g/cm3
Boiling Point 302°C
Flash Point 153°C
Packaging Amber Glass
Linear Formula C12H11N
Quantity 100 g
Sensitivity Passes Test
Solubility Information Soluble in water
Formula Weight 169.22
Grade ACS
Physical Form Solid
Chemical Name or Material Diphenylamine
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
May cause damage to organs (liver, kidneys, blood) through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Do not breathe dust.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Avoid release to the environment.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
Collect spillage.
Dispose of contents/container to comply with local, state and federal regulations.
EINECSNumber 204-539-4
Recommended Storage Room Temperature