missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Diphenylamine-4-sulfonic acid, sodium salt, pure, redox-indicator grade

CAS: 6152-67-6 | C12H11NNaO3S | 272.27 g/mol

Supplier:  Thermo Scientific Chemicals 222220250

 View more versions of this product

Catalog No. AC222220250


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Specifications

Diphenylamine-4-sulfonic acid, sodium salt
6152-67-6
Authentic
C12H11NNaO3S
25 g
(as indicator) Passes Test
sodium 4-phenylamino benzenesulfonate, sodium diphenylamine-4-sulfonate, benzenesulfonic acid, 4-phenylamino-, monosodium salt, unii-3e2ux1382o, sodium 4-anilinobenzenesulfonate, diphenylamine-4-sulfonic acid sodium salt, sodium diphenylamine-p-sulfonate, n-phenylsulfanilic acid, sodium salt, 4-phenylamino benzenesulfonic acid sodium salt, diphenylaminesulfonic acid sodium salt
VMVSDVIYIMNZBS-UHFFFAOYSA-N
4-(phenylamino)benzene-1-sulfonic acid sodium
517002
Pure
p.a.
Beige to White or Gray
Glass bottle
MFCD00036468
14, 700
15, 7423
Solubility in water: soluble
[Na].OS(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1
272.27
271.26
Crystalline Powder

Chemical Identifiers

6152-67-6
272.27
VMVSDVIYIMNZBS-UHFFFAOYSA-N
517002
C12H11NNaO3S
MFCD00036468
sodium 4-phenylamino benzenesulfonate, sodium diphenylamine-4-sulfonate, benzenesulfonic acid, 4-phenylamino-, monosodium salt, unii-3e2ux1382o, sodium 4-anilinobenzenesulfonate, diphenylamine-4-sulfonic acid sodium salt, sodium diphenylamine-p-sulfonate, n-phenylsulfanilic acid, sodium salt, 4-phenylamino benzenesulfonic acid sodium salt, diphenylaminesulfonic acid sodium salt
[Na].OS(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 228-165-6

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only