missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl selenide, 97%

Catalog No. p-7024133
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1836403 1 g
AAA1836406 5 g
AAA1836414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1836403 Supplier Thermo Scientific Chemicals Supplier No. A1836403
Only null left

CAS: 1132-39-4 | C12H10Se | 233.183 g/mol

Diphenyl selenide is used for anti-selective Michael addition of thiols and their analogs to nitro-olefins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenyl selenide is used for anti-selective Michael addition of thiols and their analogs to nitro-olefins.

Solubility
Immiscible with water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 1132-39-4
Molecular Formula C12H10Se
Molecular Weight (g/mol) 233.183
MDL Number MFCD00014067
InChI Key ORQWTLCYLDRDHK-UHFFFAOYSA-N
Synonym diphenyl selenide, diphenylselenium, phenyl selenide, biphenyl selenide, selenide, phenyl, benzene, 1,1'-selenobis, biphenyl selenium, difenylselenium, 1,1'-selenobisbenzene, diphenyl selenium
PubChem CID 14333
IUPAC Name phenylselanylbenzene
SMILES C1=CC=C(C=C1)[Se]C2=CC=CC=C2

Specifications

Melting Point 3°C
Density 1.351
Boiling Point 165°C to 167°C (12 mmHg)
Odor Unpleasant
Quantity 1 g
UN Number UN3440
Solubility Information Immiscible with water.
Formula Weight 233.17
Percent Purity 98%
Chemical Name or Material Diphenyl selenide
Hazard Category H301+H331-H373
Hazard Statement GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 214-474-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only