missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl Phthalate 98.0+%, TCI America™

Supplier:  TCI America P0305500G

 View more versions of this product

Catalog No. P0305500G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Diphenyl Phthalate
75°C
257°C
MFCD00003038
diphenyl phthalate, phenyl phthalate, phthalic acid, diphenyl ester, diphenylphthalate, 1,2-benzenedicarboxylic acid, diphenyl ester, phthalic acid diphenyl ester, caswell no. 399b, unii-bu20109xhv, 1,2-benzenedicarboxylic acid, 1,2-diphenyl ester, epa pesticide chemical code 017001
O=C(OC1=CC=CC=C1)C1=CC=CC=C1C(=O)OC1=CC=CC=C1
318.33
CHEBI:60819
≥98.0% (GC)
84-62-8
White
C20H14O4
500 g
DWNAQMUDCDVSLT-UHFFFAOYSA-N
1,2-diphenyl benzene-1,2-dicarboxylate
6778
318.33
Crystalline Powder

Chemical Identifiers

84-62-8
318.33
DWNAQMUDCDVSLT-UHFFFAOYSA-N
6778
1,2-diphenyl benzene-1,2-dicarboxylate
C20H14O4
MFCD00003038
diphenyl phthalate, phenyl phthalate, phthalic acid, diphenyl ester, diphenylphthalate, 1,2-benzenedicarboxylic acid, diphenyl ester, phthalic acid diphenyl ester, caswell no. 399b, unii-bu20109xhv, 1,2-benzenedicarboxylic acid, 1,2-diphenyl ester, epa pesticide chemical code 017001
CHEBI:60819
O=C(OC1=CC=CC=C1)C1=CC=CC=C1C(=O)OC1=CC=CC=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-3592

RTECSNumber : TI1935000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions