missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl phthalate, 98%

Catalog No. p-7021916
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1444914 25 g
AAA1444922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1444914 Supplier Thermo Scientific Chemicals Supplier No. A1444914
Only null left

CAS: 84-62-8 | C20H14O4 | 318.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 84-62-8
Molecular Formula C20H14O4
Molecular Weight (g/mol) 318.33
MDL Number MFCD00003038
InChI Key DWNAQMUDCDVSLT-UHFFFAOYSA-N
Synonym diphenyl phthalate, phenyl phthalate, phthalic acid, diphenyl ester, diphenylphthalate, 1,2-benzenedicarboxylic acid, diphenyl ester, phthalic acid diphenyl ester, caswell no. 399b, unii-bu20109xhv, 1,2-benzenedicarboxylic acid, 1,2-diphenyl ester, epa pesticide chemical code 017001
PubChem CID 6778
ChEBI CHEBI:60819
IUPAC Name diphenyl benzene-1,2-dicarboxylate
SMILES O=C(OC1=CC=CC=C1)C1=CC=CC=C1C(=O)OC1=CC=CC=C1

Specifications

Melting Point 74°C to 76°C
Density 1.28
Boiling Point 400°C to 405°C
Flash Point 224°C (435°F)
Odor Odorless
Quantity 25 g
Beilstein 2473390
Merck Index 14,7306
Formula Weight 318.33
Percent Purity 98%
Chemical Name or Material Diphenyl phthalate
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 201-546-4
RTECSNumber TI1935000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only