missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl Phosphate 99.0+%, TCI America™

Catalog No. p080125g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
25 g
Catalog No. Quantity
P080125G 25 g
P08011G 1 g
2 options

Catalog No. P080125G

Supplier: TCI America P080125G

Only null left

Chemical Identifiers

CAS 838-85-7
Molecular Formula C12H11O4P
Molecular Weight (g/mol) 250.19
MDL Number MFCD00003033
InChI Key ASMQGLCHMVWBQR-UHFFFAOYSA-N
Synonym diphenyl phosphate, phosphoric acid, diphenyl ester, phenyl hydrogen phosphate, phenyl phosphate pho 2 ho po, diphenoxyphosphinic acid, phosphoric acid diphenyl ester, diphenylphosphoric acid, phosphoric acid diphenyl, dsstox_cid_28182
PubChem CID 13282
IUPAC Name diphenoxyphosphinic acid
SMILES OP(=O)(OC1=CC=CC=C1)OC1=CC=CC=C1

Specifications

Melting Point 69°C
Color White
Quantity 25 g
Formula Weight 250.19
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Diphenyl Phosphate
RTECSNumber TC5470000
TSCA Yes